Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013887.1 | NP_001013909.1 | 6 - 7 | 800 | 96 | |
XM_006254546.2 | XP_006254608.1 | 5 - 6 | 779 | 96 | ||
GenBank mRNA | BC079289.1 | - | 6 - 7 | 800 | 7134364 | 96 |
DQ158861.1 | - | 5 - 6 | 733 | 96 | ||
FQ214241.1 | - | 5 - 6 | 653 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |