Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbfox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106974.2 | NP_001100444.2 | 2 - 3 | 559 | 285 | |
XM_006245774.2 | XP_006245836.1 | 2 - 3 | 1093 | 285 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |