Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebna1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008721.1 | NP_001008721.1 | 519 | 81 | ||
GenBank mRNA | BC087738.1 | - | 519 | 7310934 | 81 | |
DQ480746.1 | - | 1508 | 81 | |||
FQ231568.1 | - | 541 | 81 |
Gene Symbol: LOC100912375 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750007.3 | XP_003750055.1 | 4 - 5 | 539 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |