Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc48a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127456.1 | NP_001120928.1 | 1 - 2 | 185 | 95 | |
GenBank mRNA | BC158869.1 | - | 1 - 2 | 185 | 7930286 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |