Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255821.2 | XP_006255883.1 | 6 - 7 | 2834 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |