Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dfna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191749.1 | NP_001178678.1 | 8 - 9 | 1297 | 81 | |
GenBank mRNA | AY194291.1 | - | 3 - 4 | 417 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |