Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257554.2 | XP_006257616.2 | 2 - 3 | 403 | 67 | |
XM_006257555.2 | XP_006257617.1 | 2 - 3 | 426 | 67 | ||
XM_229133.8 | XP_229133.5 | 2 - 3 | 403 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |