Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257554.2 | XP_006257616.2 | 9 - 10 | 1436 | 76 | |
XM_006257555.2 | XP_006257617.1 | 9 - 10 | 1459 | 76 | ||
XM_229133.8 | XP_229133.5 | 9 - 10 | 1436 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |