Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229185.1 | XP_006229247.1 | 2 - 3 | 677 | 78 | |
XM_006229186.1 | XP_006229248.1 | 2 - 3 | 295 | 78 | ||
XM_006229187.1 | XP_006229249.1 | 2 - 3 | 248 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |