Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv39h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108883.1 | NP_001102353.1 | 4 - 5 | 1234 | 96 | |
XM_008771877.1 | XP_008770099.1 | 4 - 5 | 1364 | 96 | ||
XM_008771878.1 | XP_008770100.1 | 4 - 5 | 2438 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |