Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brdt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012031.2 | NP_001012031.1 | 2 - 3 | 327 | 116 | |
GenBank mRNA | BC078999.1 | - | 2 - 3 | 305 | 116 | |
BC158630.1 | - | 2 - 3 | 302 | 7456383 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |