Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ust | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108458.1 | NP_001101928.1 | 5 - 6 | 1062 | 118 | |
XM_008758621.1 | XP_008756843.1 | 3 - 4 | 707 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |