Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766376.1 | XP_008764598.1 | 8 - 9 | 7616 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Rattus norvegicus Probe spans exons |