Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmg20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108150.1 | NP_001101620.1 | 7 - 8 | 971 | 127 | |
XM_006243117.2 | XP_006243179.1 | 7 - 8 | 4528 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |