Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256148.1 | XP_006256210.1 | 4 - 5 | 628 | 63 | |
XM_006256149.1 | XP_006256211.1 | 4 - 5 | 619 | 63 | ||
XM_006256151.2 | XP_006256213.1 | 3 - 4 | 540 | 63 | ||
XM_006256152.2 | XP_006256214.1 | 3 - 4 | 501 | 63 | ||
XM_228042.6 | XP_228042.4 | 3 - 4 | 837 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |