Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pknox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013074.1 | NP_001013092.1 | 2 - 3 | 253 | 68 | |
XM_006256187.2 | XP_006256249.1 | 3 - 4 | 1288 | 68 | ||
XM_006256188.2 | XP_006256250.1 | 4 - 5 | 1350 | 68 | ||
XM_006256189.2 | XP_006256251.1 | 4 - 5 | 1342 | 68 | ||
XM_006256190.2 | XP_006256252.1 | 3 - 4 | 1289 | 68 | ||
XM_008772808.1 | XP_008771030.1 | 4 - 5 | 1165 | 68 | ||
XM_008772809.1 | XP_008771031.1 | 3 - 4 | 1280 | 68 | ||
XM_008772810.1 | XP_008771032.1 | 3 - 4 | 1168 | 68 | ||
GenBank mRNA | BC091397.1 | - | 2 - 3 | 253 | 7323315 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |