Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107617.2 | NP_001101087.2 | 6 - 7 | 1004 | 56 | |
XM_006256205.2 | XP_006256267.1 | 6 - 7 | 1050 | 56 | ||
XM_008772729.1 | XP_008770951.1 | 6 - 7 | 1050 | 56 | ||
GenBank mRNA | BC089841.1 | - | 6 - 7 | 971 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |