Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031832.1 | NP_114020.1 | 5 - 6 | 678 | 85 | |
GenBank mRNA | BC089054.1 | - | 5 - 6 | 666 | 7312891 | 85 |
FQ214799.1 | - | 5 - 6 | 960 | 85 | ||
FQ218337.1 | - | 5 - 6 | 678 | 85 | ||
FQ219929.1 | - | 5 - 6 | 678 | 85 | ||
FQ219987.1 | - | 5 - 6 | 678 | 85 | ||
FQ220157.1 | - | 5 - 6 | 678 | 85 | ||
FQ220174.1 | - | 5 - 6 | 677 | 85 | ||
FQ220215.1 | - | 5 - 6 | 680 | 85 | ||
FQ220224.1 | - | 5 - 6 | 672 | 85 | ||
FQ220313.1 | - | 5 - 6 | 678 | 85 | ||
FQ220316.1 | - | 6 - 7 | 908 | 85 | ||
FQ220348.1 | - | 5 - 6 | 678 | 85 | ||
FQ220361.1 | - | 5 - 6 | 837 | 85 | ||
FQ220369.1 | - | 5 - 6 | 678 | 85 | ||
FQ220439.1 | - | 5 - 6 | 677 | 85 | ||
FQ220450.1 | - | 5 - 6 | 678 | 85 | ||
FQ220455.1 | - | 6 - 7 | 905 | 85 | ||
FQ220487.1 | - | 6 - 7 | 878 | 85 | ||
FQ220505.1 | - | 5 - 6 | 678 | 85 | ||
FQ220541.1 | - | 5 - 6 | 678 | 85 | ||
FQ220563.1 | - | 5 - 6 | 678 | 85 | ||
FQ220590.1 | - | 5 - 6 | 680 | 85 | ||
FQ220652.1 | - | 5 - 6 | 678 | 85 | ||
FQ220701.1 | - | 5 - 6 | 677 | 85 | ||
FQ220837.1 | - | 5 - 6 | 677 | 85 | ||
FQ220917.1 | - | 5 - 6 | 678 | 85 | ||
FQ220938.1 | - | 5 - 6 | 678 | 85 | ||
FQ220946.1 | - | 5 - 6 | 678 | 85 | ||
FQ220952.1 | - | 5 - 6 | 678 | 85 | ||
FQ221377.1 | - | 5 - 6 | 678 | 85 | ||
FQ223467.1 | - | 5 - 6 | 680 | 85 | ||
FQ223525.1 | - | 6 - 7 | 470 | 85 | ||
FQ223531.1 | - | 5 - 6 | 678 | 85 | ||
FQ229211.1 | - | 5 - 6 | 682 | 85 | ||
FQ229259.1 | - | 5 - 6 | 678 | 85 | ||
FQ229266.1 | - | 5 - 6 | 678 | 85 | ||
FQ229277.1 | - | 5 - 6 | 675 | 85 | ||
FQ229326.1 | - | 5 - 6 | 678 | 85 | ||
FQ229348.1 | - | 5 - 6 | 678 | 85 | ||
FQ229490.1 | - | 5 - 6 | 678 | 85 | ||
FQ229494.1 | - | 5 - 6 | 678 | 85 | ||
FQ229511.1 | - | 5 - 6 | 678 | 85 | ||
FQ229560.1 | - | 5 - 6 | 680 | 85 | ||
FQ229562.1 | - | 5 - 6 | 680 | 85 | ||
FQ229564.1 | - | 5 - 6 | 680 | 85 | ||
FQ229577.1 | - | 5 - 6 | 678 | 85 | ||
FQ229661.1 | - | 5 - 6 | 678 | 85 | ||
FQ229821.1 | - | 5 - 6 | 678 | 85 | ||
FQ229967.1 | - | 5 - 6 | 680 | 85 | ||
FQ229990.1 | - | 5 - 6 | 680 | 85 | ||
FQ229991.1 | - | 5 - 6 | 680 | 85 | ||
FQ230027.1 | - | 5 - 6 | 678 | 85 | ||
FQ230050.1 | - | 5 - 6 | 681 | 85 | ||
FQ230072.1 | - | 5 - 6 | 678 | 85 | ||
FQ230121.1 | - | 5 - 6 | 685 | 85 | ||
FQ230149.1 | - | 5 - 6 | 680 | 85 | ||
J02962.1 | - | 5 - 6 | 678 | 85 | ||
KF639951.1 | - | 4 - 5 | 638 | 85 | ||
KF639952.1 | - | 4 - 5 | 638 | 85 | ||
KF639953.1 | - | 4 - 5 | 638 | 85 | ||
KF639954.1 | - | 4 - 5 | 638 | 85 | ||
KF639955.1 | - | 5 - 6 | 617 | 85 | ||
KF639956.1 | - | 5 - 6 | 617 | 85 | ||
KF639957.1 | - | 5 - 6 | 617 | 85 | ||
KF639958.1 | - | 5 - 6 | 617 | 85 | ||
M13697.1 | - | 2 - 3 | 266 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |