Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106739.1 | NP_001100209.1 | 2 - 3 | 132 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |