Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108033.1 | NP_001101503.1 | 1 - 2 | 187 | 103 | |
GenBank mRNA | BC161969.1 | - | 1 - 2 | 206 | 9028949 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |