Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr35l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099340.1 | NP_001092810.1 | 24 - 25 | 3069 | 61 | |
XM_006239885.2 | XP_006239947.1 | 25 - 26 | 3118 | 61 | ||
GenBank mRNA | EF613262.1 | - | 24 - 25 | 3069 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |