Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199083.2 | NP_954514.2 | 4 - 5 | 538 | 85 | |
GenBank mRNA | AY344057.1 | - | 4 - 5 | 538 | 85 | |
BC088091.1 | - | 4 - 5 | 515 | 7325613 | 85 | |
FQ232331.1 | - | 4 - 5 | 518 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |