Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199410.2 | NP_955442.1 | 11 - 12 | 1242 | 86 | |
GenBank mRNA | BC063180.1 | - | 11 - 12 | 1246 | 6920424 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |