Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213564.3 | NP_998729.1 | 2 | 2651 | 129 | |
XM_006256121.2 | XP_006256183.1 | 2 | 2639 | 129 | ||
XM_006256122.2 | XP_006256184.1 | 2 | 2584 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |