Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022197.2 | NP_071533.1 | 1 | 255 | 115 | |
GenBank mRNA | X06769.1 | - | 237 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Amplicon spans exons and probe does not span exons |