Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thtpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007682.1 | NP_001007683.1 | 2 | 1163 | 129 | |
GenBank mRNA | BC081978.1 | - | 2 | 1163 | 7128058 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |