Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stfa3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009177.1 | NP_001009177.1 | 1 | 56 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |