Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047853.1 | NP_001041318.1 | 3 - 4 | 319 | 89 | |
XM_006229367.2 | XP_006229429.1 | 4 - 5 | 861 | 89 | ||
XM_006229368.2 | XP_006229430.1 | 4 - 5 | 861 | 89 | ||
GenBank mRNA | AY325151.1 | - | 3 - 4 | 319 | 89 | |
AY539868.1 | - | 2 - 3 | 279 | 89 | ||
FQ222682.1 | - | 4 - 5 | 741 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons |