Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137563.1 | NP_001131035.1 | 1 - 2 | 712 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |