Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053656.2 | NP_446108.2 | 5 - 6 | 553 | 82 | |
XM_006249515.2 | XP_006249577.1 | 5 - 6 | 746 | 82 | ||
XM_006249516.2 | XP_006249578.1 | 5 - 6 | 745 | 82 | ||
XM_006249517.2 | XP_006249579.1 | 5 - 6 | 708 | 82 | ||
XM_008769306.1 | XP_008767528.1 | 5 - 6 | 422 | 82 | ||
XM_008769307.1 | XP_008767529.1 | 3 - 4 | 494 | 82 | ||
GenBank mRNA | AF020759.1 | - | 5 - 6 | 515 | 82 | |
AF064549.1 | - | 4 - 5 | 590 | 82 | ||
U14414.1 | - | 5 - 6 | 553 | 82 | ||
Y10473.1 | - | 5 - 6 | 553 | 82 | ||
Y10475.1 | - | 5 - 6 | 589 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |