Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040008.1 | NP_001035097.1 | 5 | 1446 | 73 | |
GenBank mRNA | BC079395.1 | - | 5 | 1446 | 7112099 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |