Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271026.1 | NP_001257955.1 | 2 - 3 | 199 | 107 | |
GenBank mRNA | AY387053.1 | - | 645 | 107 | ||
BC089894.1 | - | 2 - 3 | 199 | 107 | ||
BC158679.1 | - | 2 - 3 | 199 | 107 | ||
FQ227271.1 | - | 2 - 3 | 220 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |