Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271054.1 | NP_001257983.1 | 11 - 12 | 1593 | 110 | |
XM_006240151.2 | XP_006240213.1 | 11 - 12 | 1643 | 110 | ||
XM_006240152.2 | XP_006240214.1 | 12 - 13 | 1723 | 110 | ||
XM_006240153.2 | XP_006240215.1 | 11 - 12 | 2067 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |