Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271330.1 | NP_001258259.1 | 7 - 8 | 986 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |