Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276491.1 | NP_001263420.1 | 4 - 5 | 1085 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |