Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103693118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766382.1 | XP_008764604.1 | 2 - 3 | 136 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Probe spans exons |