Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001180336.1 | NP_010311.1 | 1 | 2951 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |