Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178990.3 | NP_011025.3 | 1 | 844 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |