Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244420.1 | NP_001231349.1 | 5 - 6 | 750 | 108 | |
XM_013990175.2 | XP_013845629.1 | 816 | 108 | |||
XM_013990176.2 | XP_013845630.1 | 786 | 108 | |||
GenBank mRNA | AK234343.1 | - | 5 - 6 | 750 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |