Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190232.2 | NP_001177161.2 | 3 - 4 | 342 | 82 | |
GenBank mRNA | AK230726.1 | - | 3 - 4 | 342 | 82 | |
AY233130.1 | - | 2 - 3 | 238 | 82 | ||
EW352113.2 | - | 3 - 4 | 355 | 82 | ||
HM004547.2 | - | 3 - 4 | 327 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |