Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031795.1 | NP_001026965.1 | 4 | 449 | 121 | |
XM_013988634.2 | XP_013844088.1 | 598 | 121 | |||
GenBank mRNA | AK235782.1 | - | 4 | 585 | 121 | |
AK348421.1 | - | 4 | 537 | 121 | ||
AY610421.1 | - | 4 | 525 | 121 | ||
AY996815.1 | - | 4 | 449 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |