Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRSP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213746.1 | NP_998911.1 | 5 | 552 | 133 | |
XM_005661111.3 | XP_005661168.1 | 860 | 145 | |||
XM_005661112.3 | XP_005661169.1 | 842 | 145 | |||
XM_005661113.3 | XP_005661170.1 | 674 | 145 | |||
XM_013994542.2 | XP_013849996.1 | 1041 | 145 | |||
XM_021080223.1 | XP_020935882.1 | 870 | 145 | |||
GenBank mRNA | AB114134.1 | - | 5 | 552 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |