Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213763.2 | NP_998928.1 | 4 - 5 | 356 | 131 | |
GenBank mRNA | AK234541.1 | - | 4 - 5 | 382 | 131 | |
AK391796.1 | - | 4 - 5 | 386 | 131 | ||
AK392688.1 | - | 4 - 5 | 349 | 131 | ||
AK398135.1 | - | 4 - 5 | 657 | 131 | ||
AK400566.1 | - | 4 - 5 | 578 | 131 | ||
AY459532.2 | - | 4 - 5 | 356 | 131 | ||
AY610339.1 | - | 4 - 5 | 348 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons |