Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHSR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214180.1 | NP_999345.1 | 2 | 1049 | 67 | |
XM_021068541.1 | XP_020924200.1 | 2553 | 67 | |||
XM_021068542.1 | XP_020924201.1 | 3485 | 67 | |||
GenBank mRNA | U60178.1 | - | 2 | 1049 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |