Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTNBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098600.1 | NP_001092070.1 | 9 - 10 | 861 | 96 | |
GenBank mRNA | AB271940.1 | - | 9 - 10 | 861 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |