Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100195.1 | NP_001093665.1 | 8 - 9 | 938 | 62 | |
XM_021068176.1 | XP_020923835.1 | 1178 | 62 | |||
GenBank mRNA | AK231721.1 | - | 8 - 9 | 946 | 62 | |
AK391718.1 | - | 8 - 9 | 938 | 62 | ||
AK400481.1 | - | 7 - 8 | 799 | 62 | ||
DQ351285.1 | - | 8 - 9 | 938 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |