Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105306.1 | NP_001098776.1 | 4 - 5 | 735 | 153 | |
GenBank mRNA | AK234056.1 | - | 4 - 5 | 1063 | 153 | |
AM397623.1 | - | 4 - 5 | 735 | 153 | ||
DQ859925.1 | - | 4 - 5 | 735 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |