Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123156.2 | NP_001116628.1 | 5 - 6 | 634 | 139 | |
XM_021071780.1 | XP_020927439.1 | 753 | 139 | |||
XM_021071781.1 | XP_020927440.1 | 652 | 139 | |||
GenBank mRNA | AK344387.1 | - | 5 - 6 | 668 | 139 | |
AK391517.1 | - | 5 - 6 | 635 | 139 | ||
EU402600.2 | - | 5 - 6 | 634 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |