Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001628.1 | NP_001001628.1 | 8 - 9 | 1437 | 116 | |
XM_005652441.3 | XP_005652498.1 | 2522 | 116 | |||
XM_005652443.3 | XP_005652500.1 | 2351 | 116 | |||
XM_005652444.3 | XP_005652501.1 | 2741 | 116 | |||
XM_005652445.3 | XP_005652502.1 | 2110 | 116 | |||
XM_005652446.3 | XP_005652503.1 | 1779 | 116 | |||
XM_005652447.3 | XP_005652504.1 | 1939 | 116 | |||
XM_013996333.2 | XP_013851787.1 | 3266 | 116 | |||
XM_013996365.2 | XP_013851819.1 | 1576 | 116 | |||
XM_021074699.1 | XP_020930358.1 | 3471 | 116 | |||
XM_021074701.1 | XP_020930360.1 | 882 | 116 | |||
XM_021074702.1 | XP_020930361.1 | 733 | 116 | |||
GenBank mRNA | AB370266.1 | - | 10 - 11 | 1738 | 116 | |
AK238641.1 | - | 8 - 9 | 1237 | 116 | ||
AK395023.1 | - | 9 - 10 | 1637 | 116 | ||
AY533021.1 | - | 8 - 9 | 1437 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |