Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST3GAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004047.1 | NP_001004047.1 | 5 - 6 | 946 | 57 | |
XM_005662836.3 | XP_005662893.1 | 1995 | 57 | |||
XM_005662837.3 | XP_005662894.1 | 1652 | 57 | |||
XM_005662838.3 | XP_005662895.1 | 1712 | 57 | |||
XM_013996463.2 | XP_013851917.1 | 2036 | 57 | |||
XM_021088741.1 | XP_020944400.1 | 1468 | 57 | |||
XM_021088742.1 | XP_020944401.1 | 1692 | 57 | |||
XM_021088743.1 | XP_020944402.1 | 1623 | 57 | |||
XM_021088744.1 | XP_020944403.1 | 1467 | 57 | |||
GenBank mRNA | M97753.1 | - | 5 - 6 | 946 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |